Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ATATACATACATATATATGTATATG[C/T]GTCTGTGTGTATATATACATATATG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
20 submissions
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 136132 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
FMO3 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
FMO3 - flavin containing monooxygenase 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001002294.2 | Intron | NP_001002294.1 | ||||
NM_001319173.1 | Intron | NP_001306102.1 | ||||
NM_001319174.1 | Intron | NP_001306103.1 | ||||
NM_006894.5 | Intron | NP_008825.4 | ||||
XM_011509345.2 | Intron | XP_011507647.1 |
MIR1295A - microRNA 1295a | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
MIR1295B - microRNA 1295b | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |