Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TTTCTTAGTGTTAAGGTTATTGCAG[A/G]GAGATGTTTATGGTGTATCTTTGCT
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
1 submissions
|
||||||||||||||||||||
Phenotype: |
MIM: 601013 | ||||||||||||||||||||
Literature Links: |
CACNA1E PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
CACNA1E - calcium voltage-gated channel subunit alpha1 E | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_000721.3 | Intron | NP_000712.2 | ||||
NM_001205293.1 | Intron | NP_001192222.1 | ||||
NM_001205294.1 | Intron | NP_001192223.1 | ||||
XM_017002243.1 | Intron | XP_016857732.1 | ||||
XM_017002244.1 | Intron | XP_016857733.1 | ||||
XM_017002245.1 | Intron | XP_016857734.1 | ||||
XM_017002246.1 | Intron | XP_016857735.1 | ||||
XM_017002247.1 | Intron | XP_016857736.1 | ||||
XM_017002248.1 | Intron | XP_016857737.1 | ||||
XM_017002249.1 | Intron | XP_016857738.1 | ||||
XM_017002250.1 | Intron | XP_016857739.1 | ||||
XM_017002251.1 | Intron | XP_016857740.1 |