Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CTGACCAAGGCTTCATCAAGTTTCC[C/T]GAGCCAGGTGCCATCAAAGTTCCTG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
16 submissions
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 182271 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
SPRR3 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
SPRR3 - small proline rich protein 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001097589.1 | 444 | Silent Mutation | CCC,CCT | P,P 126 | NP_001091058.1 | |
NM_005416.2 | 444 | Silent Mutation | CCC,CCT | P,P 126 | NP_005407.1 |