Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AACTCCGGCGCTGCAGCCGATCGGA[C/T]TCTGGGCCGCGGTGGGCACCGCGCG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
1 submissions
|
||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||
Literature Links: |
IGSF21 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
IGSF21 - immunoglobin superfamily member 21 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_032880.4 | 90 | UTR 5 | NP_116269.3 | |||
XM_011542319.2 | 90 | UTR 5 | XP_011540621.1 | |||
XM_017002604.1 | 90 | Intron | XP_016858093.1 | |||
XM_017002605.1 | 90 | Intron | XP_016858094.1 |