Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AGATGCCTTAAGGTGCATTCCCTGC[C/T]TCGGAGTGGCTTTGCGATTGTTTAT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
29 submissions
|
||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 603024 MIM: 610274 | ||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
ARID1A PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | ||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese)
|
||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
ARID1A - AT-rich interaction domain 1A | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
PIGV - phosphatidylinositol glycan anchor biosynthesis class V | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001202554.1 | Intron | NP_001189483.1 | ||||
NM_017837.3 | Intron | NP_060307.2 |
Set Membership: |
HapMap |