Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TAGCAATGATACCTTCCATTCTGTT[C/T]ATTTCTGGATATTTTGGCTTCAACA
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||
Literature Links: |
SESTD1 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
SESTD1 - SEC14 and spectrin domain containing 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_178123.4 | 6457 | UTR 3 | NP_835224.3 | |||
XM_011512139.2 | 6457 | UTR 3 | XP_011510441.1 | |||
XM_011512141.2 | 6457 | UTR 3 | XP_011510443.1 | |||
XM_017005250.1 | 6457 | UTR 3 | XP_016860739.1 | |||
XM_017005251.1 | 6457 | UTR 3 | XP_016860740.1 |