Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TTGTGAGAAGCGCCGGGACGCCGAG[G/T]ATGGCGCCAGCCCGGAGCAGGTTGC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 146731 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
IGFBP2 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
IGFBP2 - insulin like growth factor binding protein 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_000597.2 | 544 | Missense Mutation | GAT,TAT | D,Y 142 | NP_000588.2 | |
NM_001313990.1 | 544 | Intron | NP_001300919.1 | |||
NM_001313992.1 | 544 | Intron | NP_001300921.1 | |||
NM_001313993.1 | 544 | Intron | NP_001300922.1 |