Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CACGCTGTCACATATAACAAAATAG[C/T]TCTTGGCGGCACGGTGGCTCACGCC
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 188345 MIM: 608525 | ||||||||||||||||||||
Literature Links: |
DTYMK PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
DTYMK - deoxythymidylate kinase | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
ING5 - inhibitor of growth family member 5 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
XM_005247047.2 | Intron | XP_005247104.1 | ||||
XM_017005097.1 | Intron | XP_016860586.1 | ||||
XM_017005098.1 | Intron | XP_016860587.1 | ||||
XM_017005099.1 | Intron | XP_016860588.1 | ||||
XM_017005100.1 | Intron | XP_016860589.1 | ||||
XM_017005101.1 | Intron | XP_016860590.1 | ||||
XM_017005102.1 | Intron | XP_016860591.1 |