Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TCAACTTGAAGCTTCTCATCTTGAG[C/G]TTAATTTTACATTGCTAATGTTTGA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 609282 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
MOB1B PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
MOB1B - MOB kinase activator 1B | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001244766.1 | Intron | NP_001231695.1 | ||||
NM_001244767.1 | Intron | NP_001231696.1 | ||||
NM_173468.3 | Intron | NP_775739.1 | ||||
XM_005265709.2 | Intron | XP_005265766.1 | ||||
XM_011532412.2 | Intron | XP_011530714.1 | ||||
XM_017008837.1 | Intron | XP_016864326.1 |