Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGTGAGCCGTGTGGCAATCCAATAC[A/G]GGGGCATAGCCGGCGCTTATTTGGC
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 130590 | ||||||||||||||||||||
Literature Links: |
EEF1A1 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
EEF1A1 - eukaryotic translation elongation factor 1 alpha 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001402.5 | 1140 | Silent Mutation | CCA,CCG | P,P 359 | NP_001393.1 | |
XM_011535514.2 | 1140 | Intron | XP_011533816.1 |