Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGGAGAAATGGTGACTTTGTTTCTT[C/T]TTTTTTTTTTCCTTTCTTAACTTTG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 610045 MIM: 609269 | ||||||||||||||||||||
Literature Links: |
ALDH5A1 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
ALDH5A1 - aldehyde dehydrogenase 5 family member A1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
KIAA0319 - KIAA0319 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001168374.1 | 3976 | Intron | NP_001161846.1 | |||
NM_001168375.1 | 3976 | Intron | NP_001161847.1 | |||
NM_001168376.1 | 3976 | Intron | NP_001161848.1 | |||
NM_001168377.1 | 3976 | Intron | NP_001161849.1 | |||
NM_001252328.1 | 3976 | Intron | NP_001239257.1 | |||
NM_014809.3 | 3976 | Intron | NP_055624.2 | |||
XM_011515025.2 | 3976 | Intron | XP_011513327.1 | |||
XM_011515026.2 | 3976 | Intron | XP_011513328.1 | |||
XM_017011541.1 | 3976 | Intron | XP_016867030.1 | |||
XM_017011542.1 | 3976 | UTR 3 | XP_016867031.1 | |||
XM_017011543.1 | 3976 | Intron | XP_016867032.1 | |||
XM_017011544.1 | 3976 | Intron | XP_016867033.1 | |||
XM_017011545.1 | 3976 | Intron | XP_016867034.1 | |||
XM_017011546.1 | 3976 | UTR 3 | XP_016867035.1 | |||
XM_017011547.1 | 3976 | Intron | XP_016867036.1 | |||
XM_017011548.1 | 3976 | Intron | XP_016867037.1 | |||
XM_017011549.1 | 3976 | Intron | XP_016867038.1 | |||
XM_017011550.1 | 3976 | Intron | XP_016867039.1 | |||
XM_017011551.1 | 3976 | Intron | XP_016867040.1 | |||
XM_017011552.1 | 3976 | Intron | XP_016867041.1 | |||
XM_017011553.1 | 3976 | Intron | XP_016867042.1 |