Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AGAATAACAGGCCACAAGAGAAGAA[C/T]GCCATTTTTGACAATATCCTTTGTA
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 606857 | |||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
GCLC PubMed Links | |||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese)
|
||||||
EUR
|
||||||||
AMR
|
GCLC - glutamate-cysteine ligase catalytic subunit | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001197115.1 | 2856 | UTR 3 | NP_001184044.1 | |||
NM_001498.3 | 2856 | UTR 3 | NP_001489.1 | |||
XM_017010749.1 | 2856 | UTR 3 | XP_016866238.1 |
Set Membership: |
HapMap |