Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGAAGTAAACTGCAGAATGAAAGTC[A/G]TCTGTTCAATGTCAAAGAAGACAGG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||
Literature Links: |
TYW1B PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
TYW1B - tRNA-yW synthesizing protein 1 homolog B | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001145440.2 | Intron | NP_001138912.2 | ||||
XM_011516230.2 | Intron | XP_011514532.1 | ||||
XM_011516231.2 | Intron | XP_011514533.1 | ||||
XM_011516232.2 | Intron | XP_011514534.1 | ||||
XM_011516233.2 | Intron | XP_011514535.1 | ||||
XM_017012221.1 | Intron | XP_016867710.1 | ||||
XM_017012222.1 | Intron | XP_016867711.1 |