Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GATGCTAGGTAACAAGCGACTGGGG[C/T]TGTCCGGACTGACCCTCGCCCTGTC
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 162640 | ||||||||||||||||||||
Literature Links: |
LOC107986777 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
LOC107986777 - uncharacterized LOC107986777 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
XM_017012910.1 | 113 | Intron | XP_016868399.1 | |||
XM_017012911.1 | 113 | Intron | XP_016868400.1 |
NPY - neuropeptide Y | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_000905.3 | 113 | Silent Mutation | CTG,TTG | L,L 9 | NP_000896.1 |