Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GGCTCAACTGATCCTGCAGGCACAA[C/T]TGACCTCCGGAAGAGCCAGGCCTTG
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 612931 MIM: 606344 | |||||||||||||||||||||||||||||||||||||||||
Literature Links: |
MIR6838 PubMed Links | |||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | ||||||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | ||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese)
|
||||||
EUR
|
||||||||
AMR
|
MIR6838 - microRNA 6838 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
PGAM2 - phosphoglycerate mutase 2 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
POLM - DNA polymerase mu | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001284330.1 | 1634 | Silent Mutation | CAA,CAG | Q,Q 479 | NP_001271259.1 | |
NM_001284331.1 | 1634 | UTR 3 | NP_001271260.1 | |||
NM_013284.3 | 1634 | UTR 3 | NP_037416.1 | |||
XM_005249711.2 | 1634 | Silent Mutation | CAA,CAG | Q,Q 419 | XP_005249768.1 | |
XM_006715692.2 | 1634 | Silent Mutation | CAA,CAG | Q,Q 559 | XP_006715755.1 | |
XM_006715696.2 | 1634 | Silent Mutation | CAA,CAG | Q,Q 469 | XP_006715759.1 | |
XM_006715698.3 | 1634 | Silent Mutation | CAA,CAG | Q,Q 332 | XP_006715761.1 | |
XM_011515275.1 | 1634 | Silent Mutation | CAA,CAG | Q,Q 564 | XP_011513577.1 | |
XM_011515278.1 | 1634 | Silent Mutation | CAA,CAG | Q,Q 514 | XP_011513580.1 | |
XM_011515279.1 | 1634 | UTR 3 | XP_011513581.1 | |||
XM_011515282.2 | 1634 | Silent Mutation | CAA,CAG | Q,Q 331 | XP_011513584.1 | |
XM_011515285.1 | 1634 | Silent Mutation | CAA,CAG | Q,Q 295 | XP_011513587.1 | |
XM_011515286.2 | 1634 | Intron | XP_011513588.1 | |||
XM_011515287.2 | 1634 | Intron | XP_011513589.1 | |||
XM_017011998.1 | 1634 | Silent Mutation | CAA,CAG | Q,Q 502 | XP_016867487.1 | |
XM_017011999.1 | 1634 | Silent Mutation | CAA,CAG | Q,Q 484 | XP_016867488.1 | |
XM_017012000.1 | 1634 | UTR 3 | XP_016867489.1 | |||
XM_017012001.1 | 1634 | UTR 3 | XP_016867490.1 | |||
XM_017012002.1 | 1634 | Silent Mutation | CAA,CAG | Q,Q 295 | XP_016867491.1 | |
XM_017012003.1 | 1634 | Silent Mutation | CAA,CAG | Q,Q 295 | XP_016867492.1 | |
XM_017012004.1 | 1634 | Intron | XP_016867493.1 | |||
XM_017012005.1 | 1634 | Intron | XP_016867494.1 |
Set Membership: |
HapMap |