Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GTATCTCAGAAAGGTGCTCACCCTT[C/T]CCAAAGATAGTTGTTTTCTAATATG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||
Literature Links: |
SSPO PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
SSPO - SCO-spondin | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
ZNF862 - zinc finger protein 862 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001099220.1 | 441 | Intron | NP_001092690.1 | |||
XM_006716085.3 | 441 | Intron | XP_006716148.1 | |||
XM_011516463.1 | 441 | UTR 5 | XP_011514765.1 | |||
XM_011516464.1 | 441 | Intron | XP_011514766.1 | |||
XM_011516465.2 | 441 | Intron | XP_011514767.1 | |||
XM_011516466.2 | 441 | Intron | XP_011514768.1 | |||
XM_017012519.1 | 441 | Intron | XP_016868008.1 |