Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TTGCCGGCTCTTGCTGACAGCCCCC[A/G]CTGAGCTCCGGACCATGCAACTACA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 610334 MIM: 610385 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
LOC105375566 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
LOC105375566 - uncharacterized LOC105375566 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
TMEM176A - transmembrane protein 176A | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
TMEM176B - transmembrane protein 176B | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001101311.1 | Intron | NP_001094781.1 | ||||
NM_001101312.1 | Intron | NP_001094782.1 | ||||
NM_001101314.1 | Intron | NP_001094784.1 | ||||
NM_014020.3 | Intron | NP_054739.3 | ||||
XM_006715933.3 | Intron | XP_006715996.2 |