Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TAAATGGTAAAGTAATATGGTATCC[A/G]TTTAATTTGCCGTTTGTGCTGTCTA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 171050 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
ABCB1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
ABCB1 - ATP binding cassette subfamily B member 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_000927.4 | Intron | NP_000918.2 |
RUNDC3B - RUN domain containing 3B | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001134405.1 | Intron | NP_001127877.1 | ||||
NM_001134406.1 | Intron | NP_001127878.1 | ||||
NM_138290.2 | Intron | NP_612147.1 | ||||
XM_005250156.2 | Intron | XP_005250213.1 | ||||
XM_005250158.2 | Intron | XP_005250215.1 | ||||
XM_011515826.1 | Intron | XP_011514128.1 | ||||
XM_011515827.1 | Intron | XP_011514129.1 | ||||
XM_011515828.2 | Intron | XP_011514130.1 | ||||
XM_011515829.2 | Intron | XP_011514131.1 | ||||
XM_017011761.1 | Intron | XP_016867250.1 | ||||
XM_017011762.1 | Intron | XP_016867251.1 | ||||
XM_017011763.1 | Intron | XP_016867252.1 | ||||
XM_017011764.1 | Intron | XP_016867253.1 | ||||
XM_017011765.1 | Intron | XP_016867254.1 | ||||
XM_017011766.1 | Intron | XP_016867255.1 |