Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TATCCAGCTCAAGTACCTTCCAAAT[A/G]AGAAATTGTCAAAATCTAAATGACA
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 602447 MIM: 602720 | ||||||||||||||||||||
Literature Links: |
PON2 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
PON2 - paraoxonase 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_000305.2 | Intron | NP_000296.2 | ||||
NM_001018161.1 | Intron | NP_001018171.1 | ||||
XM_005250453.1 | Intron | XP_005250510.1 | ||||
XM_017012357.1 | Intron | XP_016867846.1 | ||||
XM_017012358.1 | Intron | XP_016867847.1 |
PON3 - paraoxonase 3 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |