Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CAAAATCCGTAGATGCCCAAGTCCC[A/G]ATATAAAATGGCATAGTATTGCATA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 158378 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
SLC20A2 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
SLC20A2 - solute carrier family 20 member 2 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
SMIM19 - small integral membrane protein 19 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001135674.1 | Intron | NP_001129146.1 | ||||
NM_001135675.1 | Intron | NP_001129147.1 | ||||
NM_001135676.1 | Intron | NP_001129148.1 | ||||
NM_138436.3 | Intron | NP_612445.2 | ||||
XM_005273398.4 | Intron | XP_005273455.1 |