Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TTCCTTTATTAGGGTAGCCCTTGCA[C/T]TTATAAAGAAACCTTCCTCAAAAAG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 604712 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
RRM2B PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
RRM2B - ribonucleotide reductase regulatory TP53 inducible subunit M2B | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001172477.1 | 4737 | UTR 3 | NP_001165948.1 | |||
NM_001172478.1 | 4737 | UTR 3 | NP_001165949.1 | |||
NM_015713.4 | 4737 | UTR 3 | NP_056528.2 |