Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TTCTAGAGAAACCTCCTTTTCCCTC[A/G]CCATTGGCTATCATGTTGCATAGTC
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 604712 | ||||||||||||||||||||
Literature Links: |
RRM2B PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
RRM2B - ribonucleotide reductase regulatory TP53 inducible subunit M2B | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001172477.1 | 2438 | UTR 3 | NP_001165948.1 | |||
NM_001172478.1 | 2438 | UTR 3 | NP_001165949.1 | |||
NM_015713.4 | 2438 | UTR 3 | NP_056528.2 |