Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CTGGGATCTTCTCTCTCTGTAGCTG[C/T]CTGTTACGTGATGACTCTTTCTTCC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 612021 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
OTUD6B PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
OTUD6B - OTU domain containing 6B | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001286745.1 | Intron | NP_001273674.1 | ||||
NM_016023.3 | Intron | NP_057107.3 | ||||
XM_011517129.2 | Intron | XP_011515431.2 |
OTUD6B-AS1 - OTUD6B antisense RNA 1 (head to head) | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |