Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ACATATAGATCAGTGAAATAGAATT[G/T]AGATTGCAGAAATAAGTGCATACAG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 602217 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
SDCBP PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
SDCBP - syndecan binding protein | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001007067.1 | Intron | NP_001007068.1 | ||||
NM_001007068.1 | Intron | NP_001007069.1 | ||||
NM_001007069.1 | Intron | NP_001007070.1 | ||||
NM_001007070.1 | Intron | NP_001007071.1 | ||||
NM_005625.3 | Intron | NP_005616.2 | ||||
XM_011517571.2 | Intron | XP_011515873.2 | ||||
XM_017013727.1 | Intron | XP_016869216.1 | ||||
XM_017013728.1 | Intron | XP_016869217.1 |