Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TCTTCAACCTTTGTACCCAGTCTGG[G/T]GGCATTTTTCTTTAGTCTCAAAATG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 604141 MIM: 611654 | ||||||||||||||||||||
Literature Links: |
ARFGEF1 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
ARFGEF1 - ADP ribosylation factor guanine nucleotide exchange factor 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_006421.4 | Intron | NP_006412.2 | ||||
XM_005251134.4 | Intron | XP_005251191.1 | ||||
XM_005251135.3 | Intron | XP_005251192.1 | ||||
XM_005251136.3 | Intron | XP_005251193.1 | ||||
XM_011517442.2 | Intron | XP_011515744.1 |