Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 603026 | |||||||||||||||||||||||||||||||||||||||||
Literature Links: |
PLAG1 PubMed Links | |||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | ||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
PLAG1 - PLAG1 zinc finger | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001114634.1 | 4427 | UTR 3 | NP_001108106.1 | |||
NM_001114635.1 | 4427 | UTR 3 | NP_001108107.1 | |||
NM_002655.2 | 4427 | UTR 3 | NP_002646.2 | |||
XM_011517544.2 | 4427 | UTR 3 | XP_011515846.1 | |||
XM_017013576.1 | 4427 | UTR 3 | XP_016869065.1 | |||
XM_017013577.1 | 4427 | UTR 3 | XP_016869066.1 |
Set Membership: |
HapMap |