Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 609774 | ||||||||||||||||||||
Literature Links: |
API5 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
API5 - apoptosis inhibitor 5 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001142930.1 | Intron | NP_001136402.1 | ||||
NM_001142931.1 | Intron | NP_001136403.1 | ||||
NM_001243747.1 | Intron | NP_001230676.1 | ||||
NM_006595.3 | Intron | NP_006586.1 | ||||
XM_006718359.3 | Intron | XP_006718422.1 | ||||
XM_017018464.1 | Intron | XP_016873953.1 | ||||
XM_017018465.1 | Intron | XP_016873954.1 |