Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GAATTTAGAGTGAATCTGGGAAATG[C/T]AGTCACCAGGTAACCCAGCAGGGCC
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 114240 | |||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
CAPN3 PubMed Links | |||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese)
|
||||||
EUR
|
||||||||
AMR
|
CAPN3 - calpain 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_000070.2 | 7731 | Intron | NP_000061.1 | |||
NM_024344.1 | 7731 | Intron | NP_077320.1 | |||
NM_173087.1 | 7731 | Intron | NP_775110.1 | |||
NM_173088.1 | 7731 | Intron | NP_775111.1 | |||
NM_173089.1 | 7731 | Intron | NP_775112.1 | |||
NM_173090.1 | 7731 | Intron | NP_775113.1 |
ZNF106 - zinc finger protein 106 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001284306.1 | 7731 | UTR 3 | NP_001271235.1 | |||
NM_001284307.1 | 7731 | UTR 3 | NP_001271236.1 | |||
NM_022473.2 | 7731 | UTR 3 | NP_071918.1 | |||
XM_005254591.2 | 7731 | UTR 3 | XP_005254648.1 | |||
XM_005254592.2 | 7731 | Intron | XP_005254649.1 | |||
XM_017022483.1 | 7731 | Intron | XP_016877972.1 | |||
XM_017022484.1 | 7731 | Intron | XP_016877973.1 | |||
XM_017022485.1 | 7731 | Intron | XP_016877974.1 |
Set Membership: |
HapMap |