Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AAAGAAAAGTTTATGTAAATGTGGG[C/T]CTAAGCACATTTATTGCAATTTCTC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 607856 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
CGNL1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
CGNL1 - cingulin like 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001252335.1 | Intron | NP_001239264.1 | ||||
NM_032866.4 | Intron | NP_116255.2 | ||||
XM_005254726.3 | Intron | XP_005254783.1 | ||||
XM_005254727.4 | Intron | XP_005254784.1 | ||||
XM_011522120.1 | Intron | XP_011520422.1 | ||||
XM_011522121.1 | Intron | XP_011520423.1 | ||||
XM_017022685.1 | Intron | XP_016878174.1 | ||||
XM_017022686.1 | Intron | XP_016878175.1 |