Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GCGGCGACCGTGACCGTGACGCGCG[A/G]GCGGGCGGCGGGGGCGCGGGCCAGG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||
Literature Links: |
DNAJA4 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
DNAJA4 - DnaJ heat shock protein family (Hsp40) member A4 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001130182.1 | 17 | UTR 5 | NP_001123654.1 | |||
NM_001130183.1 | 17 | Intron | NP_001123655.1 | |||
NM_018602.3 | 17 | Intron | NP_061072.3 | |||
XM_005254535.3 | 17 | Intron | XP_005254592.1 | |||
XM_006720601.1 | 17 | Intron | XP_006720664.1 |