Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GCACTGTGCAAAAGGCAGTATGTGC[A/G]AGGAGTGAATCAGAGCATCCGTGTG
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 118503 MIM: 118509 | |||||||||||||||||||||||||||||||||||||||||
Literature Links: |
CHRNA3 PubMed Links | |||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | ||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
CHRNA3 - cholinergic receptor nicotinic alpha 3 subunit | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
CHRNB4 - cholinergic receptor nicotinic beta 4 subunit | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_000750.3 | 2558 | Intron | NP_000741.1 | |||
NM_001256567.1 | 2558 | Intron | NP_001243496.1 | |||
XM_011521186.2 | 2558 | UTR 3 | XP_011519488.1 | |||
XM_011521187.2 | 2558 | UTR 3 | XP_011519489.1 | |||
XM_011521190.2 | 2558 | UTR 3 | XP_011519492.1 | |||
XM_011521191.2 | 2558 | UTR 3 | XP_011519493.1 | |||
XM_011521192.2 | 2558 | UTR 3 | XP_011519494.1 | |||
XM_017021885.1 | 2558 | UTR 3 | XP_016877374.1 | |||
XM_017021886.1 | 2558 | Intron | XP_016877375.1 | |||
XM_017021887.1 | 2558 | Intron | XP_016877376.1 | |||
XM_017021888.1 | 2558 | Intron | XP_016877377.1 | |||
XM_017021889.1 | 2558 | Intron | XP_016877378.1 |
Set Membership: |
HapMap |