Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AACCTAAAACAACAACAAAAACTTT[C/T]TAGGAGATTGATATGTTCATAAGAT
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 616235 | ||||||||||||||||||||
Literature Links: |
KATNBL1 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
KATNBL1 - katanin regulatory subunit B1 like 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_024713.2 | Intron | NP_078989.1 | ||||
XM_005254668.3 | Intron | XP_005254725.1 | ||||
XM_017022571.1 | Intron | XP_016878060.1 | ||||
XM_017022572.1 | Intron | XP_016878061.1 | ||||
XM_017022573.1 | Intron | XP_016878062.1 | ||||
XM_017022574.1 | Intron | XP_016878063.1 | ||||
XM_017022575.1 | Intron | XP_016878064.1 |