Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CGACAAAGCTCCTCGGCCCCAGGGC[C/T]CGCGGAGCAGACTCCGGAGCGCGGT
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 190030 MIM: 600988 | ||||||||||||||||||||
Literature Links: |
FES PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
FES - FES proto-oncogene, tyrosine kinase | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
MAN2A2 - mannosidase alpha class 2A member 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001320977.1 | 128 | UTR 5 | NP_001307906.1 | |||
NM_006122.3 | 128 | UTR 5 | NP_006113.2 | |||
XM_005254910.3 | 128 | Intron | XP_005254967.1 | |||
XM_011521565.2 | 128 | Intron | XP_011519867.1 | |||
XM_011521566.2 | 128 | Intron | XP_011519868.1 | |||
XM_017022184.1 | 128 | Intron | XP_016877673.1 | |||
XM_017022185.1 | 128 | Intron | XP_016877674.1 |