Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AGCGAATACTTTTAATGCCCGGGCC[A/G]GGAAGAAAGCGGGTTAGGATCTGAT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 185861 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
SV2B PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
SV2B - synaptic vesicle glycoprotein 2B | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001167580.1 | 140 | UTR 5 | NP_001161052.1 | |||
NM_001323031.1 | 140 | Intron | NP_001309960.1 | |||
NM_001323032.1 | 140 | UTR 5 | NP_001309961.1 | |||
NM_001323033.1 | 140 | UTR 5 | NP_001309962.1 | |||
NM_001323034.1 | 140 | UTR 5 | NP_001309963.1 | |||
NM_001323036.1 | 140 | Intron | NP_001309965.1 | |||
NM_001323037.1 | 140 | Intron | NP_001309966.1 | |||
NM_001323038.1 | 140 | Intron | NP_001309967.1 | |||
NM_001323039.1 | 140 | Intron | NP_001309968.1 | |||
NM_001323040.1 | 140 | Intron | NP_001309969.1 | |||
NM_014848.5 | 140 | UTR 5 | NP_055663.1 | |||
XM_005254998.3 | 140 | Intron | XP_005255055.1 | |||
XM_017022761.1 | 140 | Intron | XP_016878250.1 | |||
XM_017022762.1 | 140 | Intron | XP_016878251.1 |