Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GCAGCTGGCTCAGACCTTAAAACCC[C/T]TCCACGACTCATCTTTCAATATTTG
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 114835 | |||||||||||||||||||||||||||||||||||||||||
Literature Links: |
CES1 PubMed Links | |||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | ||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
CES1 - carboxylesterase 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001025194.1 | Intron | NP_001020365.1 | ||||
NM_001025195.1 | Intron | NP_001020366.1 | ||||
NM_001266.4 | Intron | NP_001257.4 | ||||
XM_005255774.1 | Intron | XP_005255831.1 |
Set Membership: |
HapMap |