Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AATTGGAGGCTGCATTGAGCCATCA[C/T]TGCACCACCGCACTTCAGCCTCAGC
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||
Literature Links: |
EFCAB3 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
EFCAB3 - EF-hand calcium binding domain 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001144933.1 | Intron | NP_001138405.1 | ||||
NM_173503.3 | Intron | NP_775774.1 | ||||
XM_011524380.1 | Intron | XP_011522682.1 | ||||
XM_011524381.1 | Intron | XP_011522683.1 |