Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGAAATCTTTTCTGTAAGTAAGCCC[A/G]TTATGAAAGATACATAGAAAATATG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 176949 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
MAPK4 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
MAPK4 - mitogen-activated protein kinase 4 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001292039.1 | Intron | NP_001278968.1 | ||||
NM_001292040.1 | Intron | NP_001278969.1 | ||||
NM_002747.3 | Intron | NP_002738.2 | ||||
XM_005258299.3 | Intron | XP_005258356.1 | ||||
XM_011526074.2 | Intron | XP_011524376.1 | ||||
XM_011526075.2 | Intron | XP_011524377.1 | ||||
XM_011526076.2 | Intron | XP_011524378.1 | ||||
XM_011526077.2 | Intron | XP_011524379.1 | ||||
XM_011526078.2 | Intron | XP_011524380.1 | ||||
XM_017025839.1 | Intron | XP_016881328.1 |