Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GGCATTACAGGCTGAGCCACTGTGC[C/T]GCCCCTCTCCTCCCTTTTGAATTAC
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 114020 | |||||||||||||||||||||||||||||||||||||||||
Literature Links: |
CDH2 PubMed Links | |||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | ||||||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | ||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese)
|
||||||
EUR
|
||||||||
AMR
|
CDH2 - cadherin 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001308176.1 | Intron | NP_001295105.1 | ||||
NM_001792.4 | Intron | NP_001783.2 | ||||
XM_011525788.1 | Intron | XP_011524090.1 | ||||
XM_017025514.1 | Intron | XP_016881003.1 |
Set Membership: |
HapMap |