Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GGAAATAAGAAGACTGAAATGGGAT[C/G]GGAAGAATAAGAATGGAAACGAAAT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 606740 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
CEACAM20 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
CEACAM20 - carcinoembryonic antigen related cell adhesion molecule 20 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001102597.2 | Intron | NP_001096067.2 | ||||
NM_001102598.2 | Intron | NP_001096068.2 | ||||
NM_001102599.2 | Intron | NP_001096069.2 | ||||
NM_001102600.2 | Intron | NP_001096070.2 | ||||
XM_011526429.2 | Intron | XP_011524731.1 | ||||
XM_011526430.2 | Intron | XP_011524732.1 | ||||
XM_011526431.2 | Intron | XP_011524733.1 | ||||
XM_011526432.2 | Intron | XP_011524734.1 |
ZNF180 - zinc finger protein 180 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |