Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TCACTTAGGTCTTCCTGTGGAGGAG[C/G]CTGGTGTGAACCGATCCAGGGACCT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 601703 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
PPM1N PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
PPM1N - protein phosphatase, Mg2+/Mn2+ dependent 1N (putative) | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
VASP - vasodilator-stimulated phosphoprotein | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_003370.3 | Intron | NP_003361.1 | ||||
XM_005259199.1 | Intron | XP_005259256.1 | ||||
XM_005259200.1 | Intron | XP_005259257.1 | ||||
XM_017027200.1 | Intron | XP_016882689.1 |