Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GGTTTGGAGAACTGGACCCTGAGGG[C/T]TGAGCTCCGATGGTGGCAAGGGGCC
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||
Literature Links: |
LOC284395 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
LOC284395 - uncharacterized LOC284395 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
VSTM2B - V-set and transmembrane domain containing 2B | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001146339.1 | Intron | NP_001139811.1 | ||||
XM_005258856.3 | Intron | XP_005258913.1 | ||||
XM_011526902.2 | Intron | XP_011525204.1 | ||||
XM_011526903.2 | Intron | XP_011525205.1 | ||||
XM_011526904.2 | Intron | XP_011525206.1 | ||||
XM_011526905.2 | Intron | XP_011525207.1 |