Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 604820 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
LILRB3 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
LILRB3 - leukocyte immunoglobulin like receptor B3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001081450.2 | 1684 | Missense Mutation | AGG,GGG | R,G 525 | NP_001074919.2 | |
NM_001320960.1 | 1684 | Missense Mutation | AGG,GGG | R,G 542 | NP_001307889.1 | |
NM_006864.3 | 1684 | Missense Mutation | AGG,GGG | R,G 525 | NP_006855.3 | |
XM_011526381.2 | 1684 | Intron | XP_011524683.1 | |||
XM_011526382.2 | 1684 | Intron | XP_011524684.1 | |||
XM_017026218.1 | 1684 | Intron | XP_016881707.1 | |||
XM_017026219.1 | 1684 | Intron | XP_016881708.1 | |||
XM_017026220.1 | 1684 | Intron | XP_016881709.1 | |||
XM_017026221.1 | 1684 | Intron | XP_016881710.1 | |||
XM_017026222.1 | 1684 | Intron | XP_016881711.1 | |||
XM_017026223.1 | 1684 | Intron | XP_016881712.1 |