Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CTCCCAGAACTCCGGACGTGTGCGG[C/T]GGTAAGCGCCCGGCCCGTACCCCCT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 114170 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
CAPNS1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
CAPNS1 - calpain small subunit 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001003962.2 | 30 | UTR 5 | NP_001003962.1 | |||
NM_001302632.1 | 30 | UTR 5 | NP_001289561.1 | |||
NM_001302633.1 | 30 | Intron | NP_001289562.1 | |||
NM_001749.3 | 30 | UTR 5 | NP_001740.1 | |||
XM_005259295.1 | 30 | UTR 5 | XP_005259352.1 | |||
XM_005259296.1 | 30 | UTR 5 | XP_005259353.1 | |||
XM_011527359.1 | 30 | Intron | XP_011525661.1 |