Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ACATTCAGGCTGGGCTCTGGATCTC[A/G]CATTCTAGCGCCCAGACATGTCGGG
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 114170 | |||||||||||||||||||||||||||||||||||||||||
Literature Links: |
CAPNS1 PubMed Links | |||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | ||||||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | ||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese)
|
||||||
EUR
|
||||||||
AMR
|
CAPNS1 - calpain small subunit 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001003962.2 | Intron | NP_001003962.1 | ||||
NM_001302632.1 | Intron | NP_001289561.1 | ||||
NM_001302633.1 | Intron | NP_001289562.1 | ||||
NM_001749.3 | Intron | NP_001740.1 | ||||
XM_005259295.1 | Intron | XP_005259352.1 | ||||
XM_005259296.1 | Intron | XP_005259353.1 | ||||
XM_011527359.1 | Intron | XP_011525661.1 |