Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GCTGCTGGGATTCGCGCTGCTGGGC[A/G]CCCACGGAGCCTCCGGGGCTGGTGA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 109480 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
BSG PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
BSG - basigin (Ok blood group) | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001322243.1 | 227 | Missense Mutation | ACC,GCC | T,A 16 | NP_001309172.1 | |
NM_001728.3 | 227 | Missense Mutation | ACC,GCC | T,A 16 | NP_001719.2 | |
NM_198589.2 | 227 | Missense Mutation | ACC,GCC | T,A 16 | NP_940991.1 | |
NM_198590.2 | 227 | Intron | NP_940992.1 | |||
NM_198591.2 | 227 | Intron | NP_940993.1 | |||
XM_017027173.1 | 227 | Missense Mutation | ACC,GCC | T,A 16 | XP_016882662.1 |