Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGGCCTCCAGGTTCACAGGCCAAAG[C/G]CCCAGGGCTCTGCCTTCCTTCCCCT
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 126375 | ||||||||||||||||||||
Literature Links: |
DNMT1 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
DNMT1 - DNA (cytosine-5-)-methyltransferase 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001130823.2 | Intron | NP_001124295.1 | ||||
NM_001318730.1 | Intron | NP_001305659.1 | ||||
NM_001318731.1 | Intron | NP_001305660.1 | ||||
NM_001379.3 | Intron | NP_001370.1 |