Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GACCTCACTCAGAGGCTCCCCCTTG[C/T]CTTAGAACCGCCCCACAGCGTGATT
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 147840 MIM: 614088 MIM: 601852 | |||||||||||||||||||||||||||||||||||||||||
Literature Links: |
ICAM1 PubMed Links | |||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | ||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
ICAM1 - intercellular adhesion molecule 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_000201.2 | Intron | NP_000192.2 |
ICAM4 - intercellular adhesion molecule 4 (Landsteiner-Wiener blood group) | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001039132.2 | Intron | NP_001034221.1 | ||||
NM_001544.4 | Intron | NP_001535.1 | ||||
NM_022377.3 | Intron | NP_071772.1 |
ICAM5 - intercellular adhesion molecule 5 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |