Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TTAAGTTTATTAAAAGCAGTTACAG[A/C]TATATTACAGATGTATTATACAGGA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 147569 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
IFNGR2 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
IFNGR2 - interferon gamma receptor 2 (interferon gamma transducer 1) | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_005534.3 | Intron | NP_005525.2 | ||||
XM_005260969.2 | Intron | XP_005261026.1 | ||||
XM_011529553.1 | Intron | XP_011527855.1 | ||||
XM_011529554.2 | Intron | XP_011527856.1 |