Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GGGAATTCACGGGTAGGAACTTCTA[C/T]AGGACACAGGTATACAGAGCTCCTT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 605413 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
RABL2B PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
RPL23AP82 - ribosomal protein L23a pseudogene 82 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |