Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AACTTTCAGTAAAATCTTGTAAACG[C/G]TTACCTTATGCCCACTCTTAGGATT
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||
Literature Links: |
SPIN3 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
SPIN3 - spindlin family member 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001010862.2 | Intron | NP_001010862.2 | ||||
XM_006724577.3 | Intron | XP_006724640.1 | ||||
XM_006724578.3 | Intron | XP_006724641.1 | ||||
XM_017029315.1 | Intron | XP_016884804.1 |